Please login to your online account to display your discounted pricing
American Educational Products Spectroscope Kit
An excellent analysis of sunlight
$237.54 - $237.54
Specifications
Age 11 to 18 years
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Product Type Price Quantity    
S53022-1A
american educational products
3093
For classroom, used to show that sunlight can be broken into colored light, and to study these colors and their relationships Spectroscope Kit Each for $237.54
Description & Specifications
Specifications
Age 11 to 18 years
This diffraction grating spectroscope is used to show that sunlight can be broken into colored light, and to study these colors and their relationships.
•    Students view different types of light and compare their spectra
•    By drawing the colors side-by-side and to the same scale, students can form their own spectrum
•    Observe and define different spectra
•    An excellent preparation for understanding methods in astronomy