Please login to your online account to display your discounted pricing
Spectronics Corporation Replacement Tubes and Filter Assembly for UV Cabinets FB-PDS-80, -81
For UV Cabinets FB-PDS-80, -81
Manufacturer: spectronics corporation  BLE-220B
 View more versions of this product
Catalog No. SPEBLE220B
  • / Each
Description & Specifications
Specifications
Wattage 4w
For Use With (Equipment) UV Cabinets FB-PDS-80, -81
Product Type Replacement Tube w/4W Filtered Tube
Wavelength 365nm