Spectroline™ Replacement Tubes
Manufacturer:  Spectroline™ BLE480B
 View more versions of this product
Catalog No. 11-992-11
Description
Description
Specifications
Specifications
365nm
E-Series Handheld lamp (EA160); Battery operated UV hand lamp
6w
Replacement Tube