Please login to your online account to display your discounted pricing
Spectroline™ Pencil Shortwave/Longwave UV Lamps
$202.49 - $202.49
Specifications
Quantity Each
Lighting Type UV pencil lamp
View More Specs
Requires:  Power supply for lamp sold separately
Products
Catalog Number Mfr. No. Diameter (English) Length (English) Wavelength Price Quantity    
11-992-54
spectronics corp
11SC2
0.25 in. 0.75 in. 254nm Each for $202.49
Description & Specifications
Specifications
Quantity Each
Lighting Type UV pencil lamp
Lightweight and miniature in size. Ideal for insertion into small apertures requiring spectroscopic wavelength calibration, sterilization, UV curing, or fluorescence detection applications. These low-pressure, mercury-vapor discharge lamps maintain a high output up to 5000 hours. Double-bore quartz construction with electrodes located at one end for easy installation.
†Relative intensity at 1 in. (2.54cm).