Please login to your online account to display your discounted pricing
Spectroline™ MiniMAX™ UV Lamps
Compact, lightweight and portable for site or field inspections
Manufacturer: spectronics corp  UV4NFW
 View more versions of this product
Catalog No. 11-992-661
  • / Each
Description & Specifications
Specifications
Wavelength Range 365 to 254nm
Wattage 4w
Quantity Each
Required Accessories 4 AA battery
Wavelength 365 nm / 254 nm/white light
Includes AC adapter, carrying strap and pouch