missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Spectroline™ Fixed Intensity TC Series Ultraviolet Transilluminators
Typical peak UV intensity of 9000 to 6000μw/cm2 (6000 to 4500μw/cm2 on dual-light models) produces brilliant fluorescence and high clarity
Manufacturer: Spectroline™ TC312R
Description
- Medium-wave 312nm UV light virtually eliminates transmission of destructive 254nm radiation
- Special diffusing screen on all 312nm and 365nm models ensures uniform light distribution for improved photographic results
- Both dual-light and variable intensity models come with a hinged UV-blocking cover and ON/OFF switches
- Variable-intensity models provide both the highest intensity and the longest sample preparation time
- Fixed-intensity models are equipped with either an ON/OFF switch or a timer that serves both as an ON/OFF switch and allows presetting of the UV exposure time
Order Info
Available in 120V 60Hz, 230V 50Hz, 240 50Hz or 100V 50/60Hz versions (specify voltage when ordering)
Specifications
9000 μw/cm2 | |
312 nm | |
34.3 cm | |
4 in. | |
18.5 in. | |
8 x 8 in. | |
Automatic shut-off timer, special diffusing screen | |
6.0 | |
20 x 20 cm | |
120/230/240/100 V 50/60 Hz | |
50/60 Hz |
UV | |
48.9 cm | |
10.16 cm | |
UV Transilluminator | |
Produce brilliant fluorescent responses and highest clarity | |
Photodoc/electronic video equipment | |
19.25 in. | |
15 w | |
19.25 x 18.5 x 4 in. (48.9 x 34.3 x 10.16cm) | |
120/230/240/100 V |