missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Spectinomycin dihydrochloride pentahydrate, Cell Culture Grade

$131.39 - $382.45

Chemical Identifiers

CAS 22189-32-8
Molecular Formula C14H36Cl2N2O12
Molecular Weight (g/mol) 495.344
MDL Number MFCD00150886
InChI Key DCHJOVNPPSBWHK-UXXUFHFZSA-N
Synonym Spectinomycin dihydrochloride
PubChem CID 67108398
ChEBI CHEBI:9217
SMILES [H+].[H+].CC1CC(=O)C2(C(O1)OC3C(C(C(C(C3O2)NC)O)NC)O)O.O.O.O.O.O.[Cl-].[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6182006 View Documents Alfa Aesar™
J6182006
5g Each for $131.39
 
AAJ6182014 View Documents Alfa Aesar™
J6182014
25g Each for $382.45
 
Description

Description

Applications
An aminocyclitol antibiotic that binds to the 30S ribosomal subunit of bacteria and interrupts protein synthesis Spectinomycin dihydrochloride pentahydrate is used as an aminocyclitol antibiotic derived from streptomyces spectabilis. It is used in cell culture applications as well as involved in the treatment of gonorrhea. Further, it is used as an anti-bacterial agent to prevent contamination and as a selection agent for resistant bacteria and plant cells.

Solubility
Soluble in water.Soluble in water, methanol, dimethyl sulfoxide, ethanol and dimethyl formamide.

Notes
Greater than 600 micrograms per milligram. Research Use only: Not intended for animal or human diagnostic or therapeutic use.Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

22189-32-8
495.344
DCHJOVNPPSBWHK-UXXUFHFZSA-N
67108398
[H+].[H+].CC1CC(=O)C2(C(O1)OC3C(C(C(C(C3O2)NC)O)NC)O)O.O.O.O.O.O.[Cl-].[Cl-]
C14H36Cl2N2O12
MFCD00150886
Spectinomycin dihydrochloride
CHEBI:9217

Specifications

22189-32-8
MFCD00150886
Soluble in water
[H+].[H+].CC1CC(=O)C2(C(O1)OC3C(C(C(C(C3O2)NC)O)NC)O)O.O.O.O.O.O.[Cl-].[Cl-]
67108398
495.35 (405.28 Anhydrous)
Cell Culture
5g
C14H36Cl2N2O12
Spectinomycin dihydrochloride
DCHJOVNPPSBWHK-UXXUFHFZSA-N
495.344
CHEBI:9217
Powder
White
Spectinomycin dihydrochloride pentahydrate
Safety and Handling

Safety and Handling

RTECSNumber : WG7400000

Recommended Storage : 2° to 8°C