Please login to your online account to display your discounted pricing

Alfa Aesar™ Spectinomycin dihydrochloride pentahydrate, 50mg/mL in water

Chemical Identifiers

Chemical Name or Material Spectinomycin dihydrochloride pentahydrate
CAS 22189-32-8
Name Note 50mg/mL in water
MDL Number MFCD00150886
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ673878EQ

 
alfa aesar™
J673878EQ
5 x 1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Spectinomycin dihydrochloride pentahydrate
Physical Form Liquid
MDL Number MFCD00150886
CAS 22189-32-8
Name Note 50mg/mL in water
Storage Note 1 -30° to -10°C
Quantity 5 x 1mL
RTECS Number WG7400000

Applications
It has been shown to have a wide variety of uses, including treating acute gonorrheal urethritis and cervictis, to mark cell layers to monitor cell fate during leaf development, as a selection marker in plant related transformation systems for plant cells containing the marker gene Spcr, to study respiratory tract infections of cattle caused by Pasteurella multocida and Mannheimia haemolytica, and to generate plants deficient for the plastid-encoded RNA polymerase on MS-agar media. Spectinomycin also has use in amplification of low copy number plasmid carrying replicons. It is recommended for use in cell culture applications at 7.5-20 mg/L.

Solubility
Fully miscible in water. Soluble in methanol, DMSO (~11 mg/ml), ethanol (~0.3 mg/ml), and DMF (~2 mg/ml).

Notes
Protect from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in freezer at -20°C.

Safety and Handling

-30° to -10°C