Please login to your online account to display your discounted pricing
Specific Heat Sets
Two specifc heat sets for demonstrations
Manufacturer: fisher science education  15-055
 View more versions of this product
Catalog No. S68852
  • / Each
Description & Specifications
Specifications
Includes Aluminum, copper, zinc, and stainless steel handy cylinders each with a machined hook for handling with tongs, instruction
Weight (English) 6 oz.
Weight (Metric) 0.17kg
Product Type Same Mass Specific Heat Set