Please login to your online account to display your discounted pricing
Specific Heat Sets
Two specifc heat sets for demonstrations
$41.10 - $41.10
Specifications
Weight (English) 6 oz.
Weight (Metric) 0.17kg
View More Specs
Products
Catalog Number Mfr. No. Includes Product Type Price Quantity    
S68852
fisher science education
15-055
Aluminum, copper, zinc, and stainless steel handy cylinders each with a machined hook for handling with tongs, instruction Same Mass Specific Heat Set Each for $41.10
Description & Specifications
Specifications
Weight (English) 6 oz.
Weight (Metric) 0.17kg
Specific Heat, Same Mass (S68852)
These four handy cylinders—aluminum, copper, zinc, and stainless steel, each with a machined hook for handling with tongs—represent all major sections of the periodic table. Same mass (58g) and diameter (19mm) for all means precise calculations of specific heat. Instructions included. Weight: 6 oz.
Specific Heat, Same Volume (S96809)
These three machined specimens were designed to fit wherever you need specimens of uniform volume. Specimens include aluminum, copper, and zinc; each measure 2 1/4 × 1 in. (6 × 2.5cm) without hooks. Instructions included. Weight: 6 oz.