missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SPC™ Universal Plus™ Sorbent Pad

Effective with hazardous and aggressive fluids

$168.12 - $168.12

Specifications

Color Green
Product Type Universal Plus Sorbent Pad
Thickness 3 ply
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Width (English) Capacity (English) Length (English) Price Quantity  
Catalog Number Mfr. No. Width (English) Capacity (English) Length (English) Price Quantity  
19-003205 Brady Canada
UN1113
12 in. 20 gal. 12 in. Case of 200 Each for $168.12
Specifications

Description

  • Chemically inert; makes pads ideal for use with aggressive fluids
  • Three-ply construction adds strength and durability yet reduces lint
  • Perforated so you use only what you need to reduces waste
  • Green color (the universal symbol for safety) indicates pads are intended for use on hazardous materials

Description & Specifications

Specifications

Green
Universal Plus Sorbent Pad
3 ply