missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SPC™ MAXX Enhanced SPC Pads

Equally effective in soaking up petroleum-based solvents or paints, vegetable oils, and other nonwater soluble chemicals

$95.12 - $106.70

Specifications

Certifications/Compliance RoHS
Color White
Density 0.04 to 0.06g/cc
Material Polypropylene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Capacity Dimensions (L x W) Price Quantity  
19033611 Brady Canada
SPC200
40 gal., 151.4L 20 x 16 in. (50.8 x 40.6cm) Case of 200 Each for $106.70
19-039-122 Brady Canada
SPC100*
35 gal., 132.4L 19 x 15 in. (48.2 x 38cm) Pack for $95.12
Specifications

Description

Sorbents do not absorb water, float indefinitely, are dustless, unaffected by temperature, flame resistant, and impervious to rot and mildew. Disposal is easy—sorbents incinerate to less than 0.02% ash.

  • SPC sorbents do not absorb water, float indefinitely, are dustless, unaffected by temperature, flame resistant, and impervious to rot and mildew
  • Disposal is easy—sorbents incinerate to less than 0.02% ash

Pads

  • Used in every oil or petroleum-based application from routine drips and leaks to major spills

Description & Specifications

Specifications

RoHS
White
0.04 to 0.06g/cc
Polypropylene