missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-Sparteine 98.0+%, TCI America™

Manufacturer:  TCI America S04611G

 View more versions of this product

Catalog No. S04611G



Specifications

Specifications

(-)-Sparteine
Undesignated
90-39-1
MFCD00069653
SLRCCWJSBJZJBV-AKRBKJBTSA-N
234.387
234.39
≥98.0% (GC)
147°C
1g
C15H26N2
Lupinidine
C1CCN2CC3CC(C2C1)CN4C3CCCC4
46937024
Clear Liquid at 20°C

Chemical Identifiers

90-39-1
234.387
SLRCCWJSBJZJBV-AKRBKJBTSA-N
46937024
C15H26N2
MFCD00069653
Lupinidine
C1CCN2CC3CC(C2C1)CN4C3CCCC4
Safety and Handling

Safety and Handling

RTECSNumber : WG5950000

TSCA : No

Recommended Storage : Refrigerator

Documents