Please login to your online account to display your discounted pricing
Whatman™ 30mm SPARTAN™ HPLC Certified Syringe Filters with Slip Luer Outlet
Provides reproducible results from the filtration of organic and aqueous solutions in HPLC. Whatman™ 30mm Spartan™ HPLC Certified Syringe Filters with Slip Luer is resistant to common HPLC solvents and exhibits very low levels of extractables that could otherwise interfere with analysis.
$289.15 - $289.15
Specifications
Diameter (Metric) 30mm
Filtration Area (Metric) 5.7cm2
Inlet Style Female luer lock
View More Specs
Products
Catalog Number Mfr. No. Pore Size Price Quantity    
09-927-425
whatman™
10462950
0.45μm Pack of 50 Each for $289.15
Description & Specifications
Specifications
Diameter (Metric) 30mm
Filtration Area (Metric) 5.7cm2
Inlet Style Female luer lock
Ready-to-use filter unit with a hydrophilic, low protein-binding membrane made of regenerated cellulose and polycarbonate housing. Tested and certified for the absence of UV-absorbing substances at wavelengths of 210 and 254nm with water, methanol and acetonitrile.
 Features
  • Excellent chemical resistance against the standard aqueous and organic HPLC solvents
  • Low dead volume (< 10μl)
Benefits
  • Versatile— Use for any application requiring a chemically resistant hydrophilic, low protein-binding membrane.
  • Documented batch-to-batch quality and consistency ensure reproducible results
  • Mini-Tip outlet is ideal for filtration into very small sample bottles
  • Purification of aqueous and organic solutions
  • Filtration of protein solutions