Please login to your online account to display your discounted pricing
Whatman™ Spartan™ HPLC Certified Syringe Filters: PROMO OFFER
Reproducible results from the filtration of organic and aqueous solutions in HPLC
$1,292.59 - $1,292.59
Specifications
Type Syringe Filter
View More Specs
Products
Catalog Number Mfr. No. Pore Size Diameter (Metric) Price Quantity    
09302151PM
whatman™
10463062PM
0.2μm 30mm Pack of 500 Each for $1,292.59
09302154PM
whatman™
10463052PM
0.45μm 30mm Pack of 500 Each for $1,292.59
Description & Specifications
Specifications
Type Syringe Filter
  • Regenerated cellulose
  • Tested for absence of UV-absorbing substances
  • Excellent chemical resistance against HPLC solvents
  • Pore size: 0.45μm