Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Modular Ultra Low Freezer Racks - PROMO
$167.20 - $167.20
Products
Catalog Number Mfr. No. Dimensions (L x W x H) Type Color Price Quantity    
14-100-152PMO
bel-art™
838874008
27 x 6 x 3.5 in.; (68.6 x 15.2 x 8.9cm) Each for $167.20
Case of 12 Each for $2,006.40
14-100-153PMO
bel-art™
838874009
27 x 6 x 3.5 in.; (68.6 x 15.2 x 8.9cm) Each for $167.20
Case of 12 Each for $2,006.40
14-100-154PMO
bel-art™
838874010
27 x 6 x 3.5 in.; (68.6 x 15.2 x 8.9cm) Each for $167.20
Case of 12 Each for $2,006.40
14-100-155PMO
bel-art™
838874011
27 x 6 x 3.5 in.; (68.6 x 15.2 x 8.9cm) Each for $167.20
Case of 12 Each for $2,006.40
Description & Specifications
Racks link togther side-by-side or can bbe securly stacked for horizontal and/or vertical storage to customize freezer space
  • Purchase as many or as few as is needed for your specific freezer space or storage needs
  • Includes pull out drawer for easy sample retrieval even from the back of the freezer
  • Each lightweight, wire formed rack holds 5 freezer boxes up to 5.25 x 5.25 x 2 in.
  • Available in 4 colors for easy sample identification
  • Optional top cover 14-100-151 provides flat shelf space for additional storage