Bel-Art™ SP Scienceware™ Earth Friendly Wide Handle Sampling Spoons
Lessen environmental impact with PLA (polylactic acid) resin sampling spoons
$29.77 - $29.77
Specifications
Bottom Shape Flat
Certifications/Compliance FDA
Product Type Earth Friendly Sampling Spoon
For Use With (Application) Designed for retrieving small, specific volume samples of powders or liquids, used as a green alternative to petroleum-based sampling tools
Handle Type Wide
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Price Quantity  
Catalog Number Mfr. No. Capacity Price Quantity  
14-512-200 Bel-Art™
F367401021
0.04 oz., 1.25mL Pack for $29.77
Case of 10 Pack for $297.70
Specifications
Description
The production of 100% plant-based PLA contributes fewer greenhouse gases and uses less nonrenewable energy from source to resin than its petroleum-based counterparts, and the plants used are an annually renewable resource. Disposal options include industrial composting for biodegradability, incineration, recycling and landfill.
  • FDA grade spoons are safe for food, drug and cosmetic contact
  • Comparable in strength and durability to polystyrene, polypropylene and other plastic resin sampling tools
  • Optimized for retrieving small, specific volume samples of powders or liquids
  • Wide handles allows user to grip firmly and provide just enough stiffness to easily sample packed powders and other firm materials
  • Flat bottom shape allows convenient duo use as weighing boats to save time and assure accuracy
  • Nonsterile
Description & Specifications
Specifications
Flat
Earth Friendly Sampling Spoon
Wide
No
FDA
Designed for retrieving small, specific volume samples of powders or liquids, used as a green alternative to petroleum-based sampling tools
PLA Resin