Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Bench-Top Biohazard Bag Holder Kit
For collecting pipette tips, microcentrifuge tubes and more, during daily use.
$33.45 - $33.45
Specifications
Autoclavable Yes
View More Specs
Includes:  100 Bench Top Biohazard Bags, made of high molecular weight, high-density (HMHD) Polyethylene and printed with Biohazard symbol and sterilization indicator patch that darkens when exposed to steam sterilization.
Products
Catalog Number Mfr. No. For Use With (Application) Product Type Price Quantity    
S04788A
bel-art™
F131660000EMD
Used to collect individual pieces of waste that need to be discarded Biohazard Bag Pack of 100 Each for $33.45
Description & Specifications
Specifications
Autoclavable Yes
  • Rectangular steel wire frame to hold one polyethylene bag of dimensions 8.5 x 11 in. (22 x 28cm)
  • Available with epoxy coat in orange color
  • Comes with snug fitting polypropylene plastic cover to hold bag securely in place
  • Reusable and allows cleaning by hand or in automated dish washers
  • Autoclavable
  • Also used with Clavies Autoclavable Bags H13182-0812
When used with lid, provides compliance with 29 CFR 1910.1030(d)(2)(xiii) (A), which requires a cover on any container used to store, transport or ship biohazardous wastes.