Please login to your online account to display your discounted pricing

Sox11 Mouse anti-Human, Biotin, Clone: SOX11-C1, eBioscience™

Mouse Monoclonal Antibody

$175.45 - $419.05

Specifications

Antigen Sox11
Clone SOX11-C1
Host Species Murine
Species Reactivity Human
Applications Immunohistochemistry (Formalin/Paraffin)
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122610

 
life technologies
LS13977380
25μg Each for $175.45

501122611

 
life technologies
LS13977382
100μg Each for $419.05
Description & Specifications

Specifications

Antigen Sox11
Clone SOX11-C1
Host Species Murine
Species Reactivity Human
Applications Immunohistochemistry (Formalin/Paraffin)
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody SOX11-C1 (C1) recognizes human Sox11. The transcription factor Sox11 is a member of the SOX (sex determining region Y-related HMG (High Mobility Group) Box) family of proteins. Under normal conditions Sox11 is expressed in the developing central nervous system during embryogenesis and has sequence homology to Sox4. Sox11 plays a role in neuronal maturation and epithelial-mesenchymal interactions and is required for the survival of neuronal and mesenchymal progenitor cells. Sox11 is expressed in mantle cell lymphoma (MCL) and in subsets of hairy cell leukemias, Burkitt lymphomas, and B cell lymphoblastic leukemias, but is not expressed in other B cell lymphomas or in normal B lymphocytes. Sox11 is also expressed in epithelial ovarian cancer and gliomas.