SouthernBiotech™ Streptavidin Reagents
Offer less nonspecific binding than egg white avidin
Manufacturer:  SouthernBiotech™ 710012
 View more versions of this product
Catalog No. OB710012
Specifications
Specifications
Light to Dark Yellow
Cyanine 3
Liquid
0.1mg
d-Biotin
0.5mg/mL
ELISA, Flow Cytometry, Immunoblotting, Immunohistochemistry
7.4
Streptomyces avidinii
2° to 8°C
Documents