Please login to your online account to display your discounted pricing
Thermo Scientific™ Sorvall™ Legend™ Micro 17 and 21 Microcentrifuge Series Promotion
Support micro-volume protocols such as nucleic acid or protein lysate preparation and PCR reaction set-up - all in a remarkably small footprint.
Specifications
Certifications/Compliance IEC 61010-1, IEC 61010-2, IEC 61010-020. CSA certified, CE marked, IVD compliant, Certified Biosafety
Capacity 24 x 1.5/2mL
Timer 1 to 99 min., ±1 min. increments
Noise Level <56dbA
Includes 24 x 1.5/2.0mL rotor
View More Specs
Products
Catalog Number Mfr. No. Electrical Requirements Type Max. RCF Max. Speed Price Quantity    
75-002-441PMO
thermo scientific™
75002441P
120V 60Hz Refrigerated 17,000 x G 13,300rpm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Certifications/Compliance IEC 61010-1, IEC 61010-2, IEC 61010-020. CSA certified, CE marked, IVD compliant, Certified Biosafety
Capacity 24 x 1.5/2mL
Timer 1 to 99 min., ±1 min. increments
Noise Level <56dbA
Includes 24 x 1.5/2.0mL rotor
Support micro-volume protocols such as nucleic acid or protein lysate preparation and PCR reaction set-up - all in a remarkably small footprint with Thermo Scientific™ Sorvall™ Legend™ Micro 17 and 21 Microcentrifuges. These microcentrifuges provide outstanding processing power, unique safety features and a broad array of rotors and accessories. Choose between 17,000 x g or 21,100 x g on both ventilated and refrigerated models.
Features:
  • Exceptional acceleration up to 21,000 × g in 12 seconds
  • Refrigerated versions offer cooling in just nine minutes
  • ClickSeal biocontainment rotor lid “clicks” in place, ensuring securely contained samples
  • New PPSU lids have improved strength and chemical resistance
  • Intuitive controls and easy to read display simplifies operation
  • Sleek, space-saving footprint
  • Rugged design for years of dependable service
  • Quite performance provides safe, stress free environment
Rotor Portfolio:
Select from one of three standard rotor configurations when purchasing a Sorvall Legend 17 or 21 microcentrifuge:
  • 24 × 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid certified by CAMR
  • 36 × 0.5mL rotor with screw-on lid
  • Dual Row rotor 18 × 2.0 /0.5mL with screw-on lid
  • PCR 8 × 8 rotor with screw-on lid
  • PCR 4 × 8 rotor with ClickSeal biocontainment lid
  • Hematocrit rotor
Ordering Information:
Biocontainment, hematocrit, PCR and dual-row rotors, lids and tube adapters are available separately.