Thermo Scientific™ Sorvall™ Legend™ 23R Fixed Angle Rotors
Effectively utilize these rotors for high-volume processing due to their 30 to 600mL capacities.
Manufacturer: Thermo Scientific™ 00555
Description
Effectively utilize Thermo Scientific™ Sorvall™ Legend™ 23R Fixed-Angle Rotors for high-volume processing due to their 30 to 600mL capacities.
- Sealable rotor 00469 can be covered with optional lid 00533 (sold separately)
- Other rotors are open-style
Ordering Information:
Tube adapters sold separately; see Thermo Sorvall Legend 23R Tube Adapters
Specifications
AM 50C.13 Angle Rotor | |
6 x 50mL | |
13,000rpm | |
Thermo Scientific™ Sorvall™ Legend™ 23R and MR23i Centrifuges |
Fixed Angle Rotor | |
21,160 x g | |
29.8mm | |
Fixed |