Thermo Scientific™ Sorvall™ Legend™ 23R Fixed Angle Rotors
Effectively utilize these rotors for high-volume processing due to their 30 to 600mL capacities.
Manufacturer:  Thermo Scientific™ 00471
 View more versions of this product
Catalog No. 00471
Description
Description
Effectively utilize Thermo Scientific™ Sorvall™ Legend™ 23R Fixed-Angle Rotors for high-volume processing due to their 30 to 600mL capacities.
  • Sealable rotor 00469 can be covered with optional lid 00533 (sold separately)
  • Other rotors are open-style
Ordering Information:
Tube adapters sold separately; see Thermo Sorvall Legend 23R Tube Adapters
Specifications
Specifications
AM 100.13 Angle Rotor
6 x 100 mL
13,000 rpm
Thermo Scientific Sorvall Legend 23R and MR23i Centrifuges
Fixed Angle Rotor
18,516 x g
38.5 mm
Fixed