Thermo Scientific™ Sorvall™ HT6 High-Capacity Floor Model Centrifuge
This product has been discontinued.
Manufacturer:  Thermo Scientific™ 11178191
Catalog No. 11-178-191
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alert:
Available in North America only.
Description
Description
This product has been discontinued. Please consider our replacement product: Thermo Scientific™ Sorvall™ RC3BP Plus Low-Speed Centrifuge.
High Throughput Processing and Application Flexibility
  • High capacity rotors with a wide selection of accessories and an unparalleled micoplate capacity offers the ability to process up to 48 standard microplates or 12 to 18 deepwell microplates in a single run
  • Processes blood and/or tissue culture tubes, microplates, clinical racks, RIA tubes, and bottles up to 1L in volume
Easy to Use
  • Intuitive VIDEOset interface provides an easy-to-read screen with online help and powerful programming features
  • 24-program memory and intuitive interface accommodates mutli-user protocols
Accurate and Dependable
  • Provides lifetime reproducibility with guaranteed run-to-run feature
  • Advanced thin film technology delivers superior sample temperature control
Unparalleled Safety
  • Built-in imbalance detection and shutdown capability
  • Rotor options that are certified for biocontainment
  • Automatic rotor recognition prevents overspeed
Convenient, Rugged and Compliant
  • Small footprint conserves lab space
  • Durable, maintenance-free brushless motor delivers years of reliable performance
Specifications
Specifications
Yes
6 x 1000 mL
-8°C to +40°C
37.2 x 30.7 x 35.4 in. (94.5 x 78 x 90 cm)
5,735 x g
94.5 cm
90 cm
30.7 in.
208 V 60 Hz
Sorvall™
Centrifuge
60 Hz
4,250 rpm
37.2 in.
35.4 in.
1 sec. to 99 min. 59 sec.; 1 min. to 99 hr. 59 min., plus HOLD
78 cm
10 acceleration/10 deceleration