missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Sorvall™ CW2 Plus Cell Washer

Simplify work and save time washing blood cells for antiglobulin reagent tests such as ABO compatibility, Rh testing, cross-matching and the Coombs procedure.



Products ${productFamilyLength}
Catalog Number Mfr. No. Description Capacity Voltage Price Quantity  
Catalog Number Mfr. No. Description Capacity Voltage Price Quantity  
36-101-9823 Thermo Scientific™
80300567
Sorvall CW2 Plus Cell Washer 12 tubes - 12mm x 75mm or 10mm x 75mm tubes 120/240V N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Simplify work and save time in washing blood cells for antiglobulin reagent tests such as ABO compatibility, Rh testing, cross-matching and the Coombs procedure with Thermo Scientific™ Sorvall™ CW2 Plus Cell Washer. The reliable choice, it is designed to support the most common applications requiring the automatic washing of donor blood.

Fully Automated for Reproducible Results

  • Automated Blood Cell Washing: Standardized, consistent procedures eliminate the variations that result from manual protocols
  • Automatic Alerts: Indicate if the saline level is low, and when a run is complete

Easy, Reliable Operation

  • Fast, Accurate Results: A time-saving wash cycle, easy to read digital displays and easy-to-use touchpad controls ensure standardized, reproducible procedures
  • Simple to Use: Simply insert the sample tubes, select the test parameters and achieve reliable results in minutes

Compact and Durable

  • Small Footprint, fits neatly on lab bench or desktop
  • Solid Construction: Low-maintenance design provides years of trouble-free use
  • Brushless motor with sealed-in lubricated bearings
  • Pump tubing is easily changed
  • High-impact, corrosion-free plastic housing

Ordering Information:

Replacement Tubing Kit (50019), Replacement Rotor (004770F), and Spare Parts Kit (with tubing, 50008) are available separately.

Includes:

Rotor (Product No. 004770F)

Recommended for:

Washing and high-speed spin for pelleting red blood cells