Please login to your online account to display your discounted pricing

Sorbitol, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™

High quality biochemicals for research use

$182.41 - $691.79

Chemical Identifiers

Chemical Name or Material Sorbitol
CAS 50-70-4
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Molecular Formula HOCH2(CHOH)4CH2OH
Formula Weight 182.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004285

 
avantor performance
V04509
Poly Pail 3kg Each for $691.79

02004286

 
avantor performance
V04507
Poly Bottle 500g Each for $182.41
Description & Specifications

Specifications

Formula Weight 182.17
CAS 50-70-4
Molecular Formula HOCH2(CHOH)4CH2OH
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Packaging Poly Pail
Quantity 3kg
Chemical Name or Material Sorbitol
Specific Gravity 1L = 1.49kg