missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sorbic Acid [for Biochemical Research], TCI America™

Manufacturer:  TCI America S08565G

 View more versions of this product

Catalog No. S08565G



Specifications

Specifications

Sorbic Acid [for Biochemical Research]
White
5g
C6H8O2
sorbic acid, 2,4-hexadienoic acid, 2e,4e-hexa-2,4-dienoic acid, 2e,4e-hexadienoic acid, panosorb, 2-propenylacrylic acid, trans,trans-sorbic acid, sorbistat, hexadienoic acid, e,e-2,4-hexadienoic acid
CC=CC=CC(=O)O
112.128
CHEBI:38358
Crystal-Powder at 20°C
228°C
134°C
110-44-1
MFCD00002703
WSWCOQWTEOXDQX-MQQKCMAXSA-N
(2E,4E)-hexa-2,4-dienoic acid
643460
112.13
≥99.0% (T)

Chemical Identifiers

110-44-1
112.128
WSWCOQWTEOXDQX-MQQKCMAXSA-N
643460
(2E,4E)-hexa-2,4-dienoic acid
C6H8O2
MFCD00002703
sorbic acid, 2,4-hexadienoic acid, 2e,4e-hexa-2,4-dienoic acid, 2e,4e-hexadienoic acid, panosorb, 2-propenylacrylic acid, trans,trans-sorbic acid, sorbistat, hexadienoic acid, e,e-2,4-hexadienoic acid
CHEBI:38358
CC=CC=CC(=O)O
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1075&(2)-0609

RTECSNumber : WG2100000

TSCA : Yes

Recommended Storage : Refrigerator

Documents