Sonoco™ Utek™ Mailing Refrigerant Packs
Phase change materials are ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or CO2
$69.76 - $140.09
Specifications
Product Type U-tek™ Refrigerant Pack
For Use With (Application) Ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or Carbon dioxide
Material Laminated, Metalized film
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Dimensions (L x W x H) Freezing Point Quantity Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W x H) Freezing Point Quantity Price Quantity  
03-528-4 Sonoco™
TS432
6 x 10.6 x 0.75 in. (15.2 x 2.5 x 1.9cm) -23°C 36 Case Case of 36 Each for $140.09
03-525-7F Sonoco™
TS426
10.5 x 7.75 x 1.18 in. (26.7 x 19.5 x 2.9cm) -1°C 6 Case Case of 6 Each for $69.76
03-525-7C Sonoco™
TS597
6.5 x 10.25 x 0.87 in. (16.5 x 10.8 x 2.2cm) -23°C 16 Case Case of 16 Each for $77.24
Specifications
Description
  • Gel packs have a laminated, metalized film exterior that provides radiant heat barrier, resists punctures
  • Single packets are ideal for use as a cubed ice substitute
  • Mat is well suited for wraparound applications
Description & Specifications
Specifications
U-tek™ Refrigerant Pack
Ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or Carbon dioxide
Laminated, Metalized film