Sonoco™ ThermoSafe Reusable Foam-Only EPS Bio-Mailers
EPS foam-only shippers offer solution for safe storage and delivery of most refrigerated or frozen products
$362.38 - $362.38
Specifications
Material EPS
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Dimensions (L x W x H) Exterior Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W x H) Exterior Price Quantity  
03-525-5 Sonoco™
TS445*
Case for $362.38
Specifications
Description
  • Reusable
  • Large selection of cost-effective, reusable stock containers available
  • Excellent dry ice sublimation – average dry ice usage in 1 1/2 in. thick wall units is 5 pounds over 24 hours, and 3-4 pounds thereafter
Description & Specifications
Specifications
EPS