Sonoco™ ThermoSafe Reusable EPS Foam Stock Containers
EPS foam-only shippers offer solution for safe storage and delivery of most refrigerated or frozen products
$62.59 - $246.38
Specifications
Material Reusable EPS foam
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Dimensions (L x W x H) Exterior Quantity Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W x H) Exterior Quantity Price Quantity  
03-001-224 Sonoco™
TS809
21 x 21 x 16.75 in. (53.3 x 53.3 x 42.5cm) 2/Cs. Case of 2 Each for $246.38
03-001-223 Sonoco™
TS428
20.25 x 13.25 x 13.75 in. (51.4 x 33.6 x 34.9cm) 4/Cs. Case of 4 Each for $242.38
03-001-227 Sonoco™
TS673
17.62 x 13.37 x 9.6 in. (44.7 x 33.9 x 24.3cm) 2/Cs. Case of 2 Each for $62.59
Specifications
Description
  • Reusable
  • Large selection of cost-effective containers available
  • Excellent dry ice sublimation – average dry ice usage in 1 1/2 in. thick wall units is 5 pounds over 24 hours, and 3-4 pounds thereafter
Description & Specifications
Specifications
Reusable EPS foam