Sonoco™ ThermoSafe Nylon Carriers
Rugged lightweight totes are ideal for transport of frozen or refrigerated food and medical products
$223.18 - $236.56
Specifications
Material Nylon shell; Expanded Polystyrene (EPS) foam insulation
Dimensions (L x W x H) Exterior 37.7 x 33.3 x 30.8cm (14.88 x 13.13 x 12.13 in.)
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
03-525-60 Sonoco™
TS486RED
Red Each for $223.18
03-525-61 Sonoco™
TS486BLUE
Blue Each for $236.56
Specifications
Description
  • Washable
  • Weather-resistant nylon shell with thick-walled EPS foam insulation
  • Adjustable nylon straps for hand or shoulder carrying
  • 0.59 cu. ft.
Description & Specifications
Specifications
Nylon shell; Expanded Polystyrene (EPS) foam insulation
37.7 x 33.3 x 30.8cm (14.88 x 13.13 x 12.13 in.)