Sonoco™ ThermoSafe Nomadic™ Pre-Qualified Shippers
For protection of refrigerated and frozen pharmaceuticals and biological samples and specimens
Manufacturer:  Sonoco™ TSE24A
 View more versions of this product
Catalog No. 03-600-122
Specifications Description & Specifications
Specifications
Polyurethene (PUR) Foam
15.5 in.
39.3cm
15.9 in.
40.3cm
15.9 in.
40.3cm
For refrigerated, frozen pharmaceutical and biological specimens
−2 to 8°C
24 hours
3.7 in.
9.3cm
7.5 in.
19cm
5 in.
12.7cm
15.9 x 15.9 x 15.5 in. (40.3 x 40.3 x 39.3cm)