Please login to your online account to display your discounted pricing
Sonoco™ ThermoSafe Multi-Purpose Foam Container Assembled in Corrugated Carton
EPS foam-only shippers offer solution for safe storage and delivery of most refrigerated or frozen products
$182.74 - $460.21
Specifications
Material Expanded Polystyrene (EPS) Insulated
View More Specs
Products
Catalog Number Mfr. No. Dimensions (L x W x H) Exterior Quantity Price Quantity    
03-528-12
sonoco™
TS638
28.25 x 17.25 x 15.625 in. (71.755 x 43.815 x 39.6875 cm) 2/Cs. Case of 2 Each for $182.74
03-530-79
sonoco™
TS609UPS
9 x 7.5 x 8 in. (22. 86 x 19.05 x 20.32 cm) 24/Cs. Case of 24 Each for $460.21
Description & Specifications
Specifications
Material Expanded Polystyrene (EPS) Insulated
  • Large selection of cost-effective, reusable stock containers available
  • Excellent dry ice sublimation – average dry ice usage in 1 1/2 in. thick wall units is 5 pounds over 24 hours, and 3-4 pounds thereafter