Please login to your online account to display your discounted pricing

Sonoco™ ThermoSafe LabMailer Sample-Guard 28mL Vial Shippers

Insulated shippers with lid eliminate sample leakage and breakage

$442.97 - $568.17

Specifications

Includes One 24 oz. Phase Change Material Reusable Pack
Material Insulated Expanded Polystyrene (EPS) Foam
View More Specs
Includes:  One 24 oz. U-tek™ phase change material, Reusable polyethylene platform
Products
Catalog Number Mfr. No. Quantity Price Quantity    

03-528-11

 
sonoco™
TS480413
12 x 40mL Vials Case of 8 Each for $442.97

11-675-47

 
sonoco™
TS480490
48 x 40mL Vials Case of 8 Each for $568.17
Description & Specifications

Specifications

Includes One 24 oz. Phase Change Material Reusable Pack
Material Insulated Expanded Polystyrene (EPS) Foam

  • Expanded polystyrene foam shippers secure upright twelve or forty-eight 28mL vials
  • Shippers maintain required sample temperature for 72 hours or longer
  • Shippers are assembled in 200 lb. test corrugated cartons

Weight empty includes refrigerant.