Sonoco™ ThermoSafe Infectious Shipper
Manufacturer:  Sonoco™ 630
 View more versions of this product
Catalog No. 03-528-32
Specifications Description & Specifications
Specifications
High-density Expanded Polystyrene (PS)
Meets the specifications for testing and performance by D.O.T. Regulation 49 CFR Parts 178 to 179 and ICAO Technical Instructions, Chapter 6.
Foam container molded of high-density expanded polystyreneAbsorbent pad; Screw cap aluminum can measuring 3D x 6 in.H (7.6 x 15cm); Flexible foam insert with four pre-drilled holes to accommodate supplied test tubes; Four 10mL sterile blood collection