Please login to your online account to display your discounted pricing
Sonoco™ ThermoSafe™ GREENBOX™ Thermal Management Systems: CRT (15-30°C)
GREENBOX is a sustainable packaging, designed for the safe transportation of temperature sensitive products.
$699.73 - $1,992.68
Specifications
Length (English) Exterior 9 in.
Width (English) Exterior 9 in.
Height (English) Exterior 9 in.
Material Vacuum Insulated Panels (VIP)
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Payload Price Quantity    
03-531-241
sonoco™
TS09UC3
Each for $699.73
03-531-242
sonoco™
TS09UC5
Each for $752.03
03-531-248
sonoco™
TS24UC6
18.63L Each for $1,992.68
Description & Specifications
Specifications
Length (English) Exterior 9 in.
Width (English) Exterior 9 in.
Height (English) Exterior 9 in.
Material Vacuum Insulated Panels (VIP)
Greenbox™ Reusable Insulated Shippers have been designed to utilize simple packaging configurations, reduce operational
complexity, and optimize total cost of ownership. GreenBoxs simple design and modular format increases reclamation and
refurbishment efficiencies. Ensure the efficacy of your valuable temperature-sensitive products and maintain regulatory
compliance throughout distribution with Greenbox.
  • ZERO Bench-Time™ refrigerant design means no waiting, greatly simplifies and accelerates the assembly process
  • Utilizes VIP insulation and PureTemp™ PCMs resulting in fewer components and lower weight, decreasing total cost of ownership