Sonoco™ ThermoSafe Foam Shipper for 48 hrs and more Shipments
EPS foam-only shippers offer solution for safe storage and delivery of most refrigerated or frozen products
$223.06 - $327.59
Specifications
For Use With Shipments 48 hrs and more
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Dimensions (L x W x H) Exterior Quantity Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W x H) Exterior Quantity Price Quantity  
03-001-120 Sonoco™
TS651
11 x 9 x 10 in. (28 x 22.8 x 25.4cm) 12 Case of 12 Each for $223.06
03-001-133 Sonoco™
TSD646
8 x 6.5 x 4.5 in. (20.3 x 16.5 x 11.4cm) 36 Case of 36 Each for $327.59
Specifications
Description
  • Large selection of cost-effective containers available
  • Excellent dry ice sublimation – average dry ice usage in 1 1/2 in. thick wall units is 5 pounds over 24 hours, and 3-4 pounds thereafter
Description & Specifications
Specifications
Shipments 48 hrs and more