Please login to your online account to display your discounted pricing
Sonoco™ ThermoSafe Foam-Only EPS Insulated Shippers
EPS foam-only shippers offer solution for safe storage and delivery of most refrigerated or frozen products
$247.76 - $302.59
Specifications
Material Expanded Polystyrene (EPS) Insulated
View More Specs
Products
Catalog Number Mfr. No. Dimensions (L x W x H) Exterior Quantity Price Quantity    
03-530-84
sonoco™
TS621UPS
Case of 8 Each for $289.68
03-530-50
sonoco™
TS315UPS
Case of 6 Each for $247.76
03-530-78
sonoco™
608UPS
Case of 27 Each for $302.59
Description & Specifications
Specifications
Material Expanded Polystyrene (EPS) Insulated
  • Large selection of cost-effective, reusable stock containers available
  • Excellent dry ice sublimation – average dry ice usage in 11/2 in. thick wall units is 5 pounds over 24 hours, and 3-4 pounds thereafter