Sonoco™ ThermoSafe Envoy™ Global Pre-Qualified Insulated Shippers with Winter Packout
Temperature controlled packaging solution
Manufacturer:  Sonoco™ TSE24R2W
 View more versions of this product
Catalog No. 03-531-707
Specifications Description & Specifications
Specifications
Expanded Polystyrene
44 lb.
4 Pallets
24.1L
Refrigerated: 2-8°C
6.36 gal.
20kg
737 x 521 x 540mm (29 x 20.5 x 21.26 in.)
Insulated Shipper with Winter Packout Configuration