Sonoco™ ThermoSafe Envoy™ Global Pre-Qualified Insulated Shippers: Summer Packout
Temperature controlled packaging solution.
Manufacturer:  Sonoco™ TSE8C2S
 View more versions of this product
Catalog No. 03-531-710
Specifications Description & Specifications
Specifications
Expanded polystyrene
21.2 in.
16.7 in.
19.2 lb.
15.7 in.
12 Pallets
8.1L
CRT: 15-25°C
2.13 gal.
540mm
425mm
8.7kg
400mm
424 x 400 x 540mm (16.7 x 15.75 x 21.26 in.)
Insulated Shipper with Summer Packout Configuration