Please login to your online account to display your discounted pricing

Sonoco™ ThermoSafe Certis™ Gold Shipper Box with Universal Packout


Accommodates broad range of temperature-sensitive products

Manufacturer: sonoco™  TSCRUG3XP

 View more versions of this product

Catalog No. 03-531-201

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

For Use With Accommodates broad range of temperature-sensitive products
Nominal Payload (English) 40 lb.
Color Gold
Elapsed Time Range 78hrs.
Material Polyurethane (PUR)