Sonoco™ PureTemp™ Phase Change Materials
PureTemp™ Phase Change Materials liquefy or solidify at precise temperatures. PureTemp comes in foam bricks and hard bottles.
$755.21 - $1,422.29
Specifications
Product Type Phase Change Material
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Dimensions (L x W x H) Quantity Temperature (Metric) Phase Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W x H) Quantity Temperature (Metric) Phase Price Quantity  
03-600-110 Sonoco™
TSTPB3000Y
10.75 x 4.25 x 0.5 in. (27.3 x 29 x 1.27cm) 24 Case 20°C Case of 24 Each for $1,243.88
03-600-108 Sonoco™
TSTPB2000A
9 x 3 x 0.5 in. (22.8 x 16.5 x 1.27cm) 48 Case 4°C Case of 48 Each for $1,200.15
03-600-109 Sonoco™
TSTPB3000A
10.75 x 18 x 0.5 in. (27.3 x 29 x 1.27cm) 24 Case 4°C Case of 24 Each for $1,343.68
03-600-111 Sonoco™
TSB27A
8 x 4.5 x 1 in. (20.32 x 13.9 x 2.54cm) 16 Case 4°C Case of 16 Each for $1,279.44
03-600-112 Sonoco™
TSB27G
8 x 0.75 x 1 in. (20.32 x 0.39 x 2.54cm) 16 Case 1°C Case of 16 Each for $824.65
03-600-113 Sonoco™
TSB27R
8 x 8 x 1 in. (20.32 x 29 x 2.54cm) 16 Case 23°C Case of 16 Each for $1,366.91
03-600-114 Sonoco™
TSB38A
10 x 2 x 1 in. (25 x 15.8 x 2.54cm) 12 Case 4°C Case of 12 Each for $1,302.00
03-600-115 Sonoco™
TSB38G
10 x 12.7 x 1 in. (25 x 30 x 2.54cm) 12 Case 1°C Case of 12 Each for $755.21
03-600-116 Sonoco™
TSB38R
10 x 11 x 1 in. (25 x 26.03 x 2.54cm) 12 Case 23°C Case of 12 Each for $1,422.29
Specifications
Description
PureTemp Phase Change Materials liquefy or solidify at precise temperatures. When the surrounding temperature falls below the phase temperature, they solidify and release their stored energy. If the temperature rises above the phase temperature, they liquefy and absorb excess energy. 
These two phenomena protect the product from excessive heat and cold to help maintain product temperature for extended periods of time. PureTemp is engineered to react to a precise temperature, from -40° to +37°C, and to operate in a very narrow temperature range. It is also renewable and biodegradable.
PureTemp comes in foam bricks and hard bottles.
SUSTAINABILITY
  • Biodegradable
  • USDA BioPreferred™ designation
TIGHTER TEMPERATURE CONTROL
  • Extensive range of temperature options
  • Constant temperature during freezing or melting
  • provides constant temperature control
  • Eliminates thermal shock with fewer excursions
TOTAL COST CONTROL
  • Increases payload efficiency
  • Reduces shipping weight and freight costs. 
  • Less PureTemp material is required for temperature control when compared to water-based refrigerants
  • Simplifies packout inventory and process
  • Decreases box size for less warehouse and packout space
  • Eliminates bench-time
  • Increases shipping duration
Description & Specifications
Specifications
Phase Change Material