Please login to your online account to display your discounted pricing

Somatostatin Mouse, eFluor 660, Clone: ICDCLS, eBioscience™

Mouse Monoclonal Antibody

$146.45 - $397.30

Specifications

Antigen Somatostatin
Clone ICDCLS
Host Species Mouse
Gene Alias SST
Species Reactivity Human, Mouse, Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124527

 
life technologies
LS50975180
25μg Each for $146.45

501124528

 
life technologies
LS50975182
100μg Each for $397.30
Description & Specifications

Specifications

Antigen Somatostatin
Clone ICDCLS
Host Species Mouse
Gene Alias SST
Species Reactivity Human, Mouse, Rat
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Microscopy
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody ICDCLS recognizes human, mouse, and rat Somatostatin. Somatostatin, also known as growth hormone-inhibiting hormone (GHIH) or somatotrophin release-inhibiting factor (SRIF), is a peptide hormone with two active forms (14 and 28 amino acids) produced by alternative cleavage of a single preproprotein. Somatostatin is secreted by the delta cells of the islet of Langerhans in the pancreas as well as by the hypothalamus and intestine. Somatostatin has a variety of functions, such as regulating the endocrine system and affecting neurotransmission and cell proliferation. Somatostatin also inhibits the release of numerous secondary hormones, such as insulin and glucagon. Somatostatin can be used to identify medium size aspiny neurons in the striatum, cortex, and hippocampus. Expression of somatostatin is present in hyperplasia of the pancreatic islets as well as islet cell tumors such as somatostatinomas.