missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Solketal, 97%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 153651000

 View more versions of this product

Catalog No. AC153651000



Specifications

Specifications

Solketal
90°C
Glass bottle
1.06
188.0°C to 189.0°C
-50°C
97%
96% min. (GC)
MFCD00063238
15, 5259
Solubility in water: miscible. Other solubilities: miscible with alcohol, acetals, esters, ethers,, aromatic hydrocarbons, chlorinated hydrocarbons,, gasolines and petr.ether
CC1(OCC(O1)CO)C
132.16
132.16
97%
1.0600g/mL
Authentic
1.4330 to 1.4350
11 mPa.s (20°C)
Colorless
100g
100-79-8
C6H12O3
19, V,2, 362
1,2-isopropylideneglycerol
RNVYQYLELCKWAN-UHFFFAOYSA-N
(2,2-dimethyl-1,3-dioxolan-4-yl)methanol
7528
Liquid

Chemical Identifiers

100-79-8
132.16
RNVYQYLELCKWAN-UHFFFAOYSA-N
7528
CC1(OCC(O1)CO)C
C6H12O3
MFCD00063238
1,2-isopropylideneglycerol
(2,2-dimethyl-1,3-dioxolan-4-yl)methanol
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 202-888-7

RTECSNumber : JI0400000

TSCA : TSCA

SDS Documents