Please login to your online account to display your discounted pricing
Solid-State Induction Coils
Manufacturer: fisher science education  37733
 View more versions of this product
Catalog No. S43520
  • / Each
Description & Specifications
Specifications
Voltage 115V
For Use With (Application) Limits current and extends the life of the vacuum discharge tubes
Product Type Induction Coil, 3 in. Spark
This Solid-State Induction Coil limits current and extends the life of the vacuum-discharge tubes it powers. Both models operate off a 115V, 60Hz line. The oscillator drives the coil at adjustable voltages with corresponding currents.