Please login to your online account to display your discounted pricing
Solid-State Induction Coils
$792.00 - $792.00
Specifications
For Use With (Application) Limits current and extends the life of the vacuum discharge tubes
View More Specs
Products
Catalog Number Mfr. No. Product Type Voltage Price Quantity    
S43520
fisher science education
37733
Induction Coil, 3 in. Spark 115V Each for $792.00
Description & Specifications
Specifications
For Use With (Application) Limits current and extends the life of the vacuum discharge tubes
This Solid-State Induction Coil limits current and extends the life of the vacuum-discharge tubes it powers. Both models operate off a 115V, 60Hz line. The oscillator drives the coil at adjustable voltages with corresponding currents.