Please login to your online account to display your discounted pricing

Moldex™ Softies™ Foam Ear Plugs

Soft comfortable fit and high protection

$243.53 - $243.53

Specifications

Certifications/Compliance ANSI S3.19-1974
Color Orange
Product Type Softies™ Earplug
Size One size fits to all
For Use With (Application) Extra soft, low pressure Foam for comfort and fit
View More Specs
Products
Catalog Number Mfr. No. Style Price Quantity    

19-813-682

 
moldex™
6600**
Uncorded Pack of 200 Pair for $243.53
Description & Specifications

Specifications

Certifications/Compliance ANSI S3.19-1974
Color Orange
Product Type Softies™ Earplug
Size One size fits to all
For Use With (Application) Extra soft, low pressure Foam for comfort and fit
Material Foam
Noise Reduction Range (NRR) 33dB
Shape Tapered

  • Extra soft foam and tapered shape makes for a gentle and comfortable seal
  • Radiant orange color makes compliance checks easy
  • Fits even very small ear canals
  • PVC free
  • Noise Reduction Rating: 33