Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium zirconium oxide, 98%

Chemical Identifiers

Chemical Name or Material Sodium zirconium oxide
CAS 12201-48-8
Formula Weight 185.2
MDL Number MFCD00014245
Synonym Sodium zirconate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1876214

 
alfa aesar™
1876214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1876222

 
alfa aesar™
1876222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 185.2
Chemical Name or Material Sodium zirconium oxide
TSCA TSCA
Mesh Size −100 Mesh
Physical Form Powder
Assay 98%
MDL Number MFCD00014245
CAS 12201-48-8
Synonym Sodium zirconate
Color White
Quantity 25g