missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium tungsten oxide dihydrate, 95%

Manufacturer:  Alfa Aesar™ 8737130

 View more versions of this product

Catalog No. AA8737130



Specifications

Specifications

Sodium tungsten oxide dihydrate
H4Na2O6W
14,8698
Soluble in water; Insoluble in alcohol
O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]
329.846
CHEBI:63939
Crystalline
3.250g/mL
White
250g
10213-10-2
MFCD00149190
sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
WPZFLQRLSGVIAA-UHFFFAOYSA-N
disodium;dioxido(dioxo)tungsten;dihydrate
150191
329.86 (293.83 Anhydrous)
95%
Odorless
100°C (in -2H2O)

Chemical Identifiers

10213-10-2
329.846
WPZFLQRLSGVIAA-UHFFFAOYSA-N
150191
disodium;dioxido(dioxo)tungsten;dihydrate
H4Na2O6W
MFCD00149190
sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
CHEBI:63939
O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]
Safety and Handling

Safety and Handling

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : YO7900000

TSCA : TSCA

SDS Documents