Learn More
Sodium tungstate dihydrate, 99+%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 207625000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent for analysisSpecifications
Sodium tungstate dihydrate | |
10213-10-2 | |
Crystalline Flakes or Powder | |
99% min | |
Na2WO4·2H2O | |
15,295 | |
sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o | |
WPZFLQRLSGVIAA-UHFFFAOYSA-N | |
disodium;dioxido(dioxo)tungsten;dihydrate | |
150191 | |
329.85 | |
Analytical | |
4.18 |
p.a. | |
White | |
500 g | |
Na2O4W·2H2O | |
MFCD00149190 | |
15,8826 | |
Solubility in water: 730g/L (20°C). Other solubilities: insoluble in alcohol and acids | |
O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+] | |
329.85 | |
CHEBI:63939 | |
≥99% | |
Glass bottle |
Chemical Identifiers
10213-10-2 | |
329.85 | |
WPZFLQRLSGVIAA-UHFFFAOYSA-N | |
150191 | |
disodium;dioxido(dioxo)tungsten;dihydrate |
Na2O4W·2H2O | |
MFCD00149190 | |
sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o | |
CHEBI:63939 | |
O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+] |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove
GHS Signal Word: Warning
RTECSNumber : YO7900000
RUO – Research Use Only